

(R)-(+)-3-Methoxy-α-methylbenzylamine ≥99%, ee 98%, ChiPros®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:88196-70-7
:
Formula:C₉H₁₃NO
MDL Number:MFCD00671655
:151.21 g/mol
Boiling Pt:66 °C (0.38 mmHg)
Melting Pt:5 °C
Density:1.023
Flash Pt:>100 °C (212 °F)
Storage Temperature:Ambient
UN:2735
ADR:8,III
:GHS05
:GHS07
Related Information
Frequently Bought Together