
Lidocaine 97.5%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:137-58-6
:
Formula:C₁₄H₂₂N₂O
MDL Number:MFCD00026733
Boiling Pt:182 °C (4 mmHg)
Melting Pt:66…69 °C
:234.34 g/mol
Storage Temperature:Ambient
EINECS:205-302-8
:GHS07
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:66 to 69 °C
- Titration with HClO4:≥96.5 %
- Specific optical rotation:-8° to -12° (20 °C, 589 nm) (c=1, CH3OH)
Specification Test Results
Frequently Bought Together