Farnesol (mixture of isomers) 96%
:4602-84-0
:
:C₁₅H₂₆O
:MFCD00002918
:222.37 g/mol
:149 °C (4 mmHg)
:0.8865 g/cm³
:96 °C
:Ambient
:225-004-1
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4870 to 1.4920 (20°C, 589 nm)
- Density:(20/4°C) 0.884 to 0.89 kg/l
- GC:≥95 % (sum of geometric isomers)
Frequently Bought Together