Scheide- und Tropftrichter-Rohkörper, DURAN®
Supplier: DWK Life Sciences
Certificates
About this item
DURAN®, borosilicate glass 3.3, clear.
- cylindrical with stopper bed for ISO 4800
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
For glass manipulators.
Frequently Bought Together