Order Entry
Austria
ContactUsLinkComponent
Toluene, anhydrous (max. 0.002% H₂O) ≥99.8%
Flammable
Health
Irritant
Toluene, anhydrous (max. 0.002% H₂O) ≥99.8%
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Health
Irritant
Toluene, anhydrous (max. 0.002% H₂O) ≥99.8%
Supplier:  VWR Chemicals
Certificates
Limited Time Sale
Select items below are currently on sale

Specification Test Results

  • Assay (calculated on anhydrous):
    Min. 99.8 %
  • Acidity:
    Max. 0.0005 meq/g
  • Residue on evaporation:
    Max. 0.0005 %
  • Water (K.F.):
    Max. 0.0020 %

Specification Test Results

Related Information

Frequently Bought Together