Order Entry
Austria
ContactUsLinkComponent
Toluene ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Flammable
Health
Irritant
Toluene ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Health
Irritant
Toluene ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (GC):
    Min. 99,8 %
  • Acidity:
    Max. 0.0005 meq/g
  • Alkalinity:
    Max. 0.0002 meq/g
  • Evaporation residue:
    Max. 5 ppm
  • Water:
    Max. 0.05 %
  • Absorbance (300 nm):
    Max. 0.15
  • Absorbance (310 nm):
    Max. 0.1
  • Absorbance (330 nm):
    Max. 0.02
  • Absorbance (350 nm):
    Max. 0.01
  • Transmittance (300 nm):
    Min. 70 %
  • Transmittance (310 nm):
    Min. 80 %
  • Transmittance (330 nm):
    Min. 95 %
  • Transmittance (350 nm):
    Min. 98 %

Specification Test Results

Frequently Bought Together