Order Entry
Austria
ContactUsLinkComponent
Toluene ≥99%, GPR RECTAPUR® for pathology
Flammable
Health
Irritant
Toluene ≥99%, GPR RECTAPUR® for pathology
Catalog # 28684.364
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Health
Irritant
Toluene ≥99%, GPR RECTAPUR® for pathology
Catalog # 28684.364
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle, fluorinated
  • Pk:
    5 L
  • Seq:
    5190

Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Boiling point:
    109 - 112 °C
  • Density (20/4):
    0.865 - 0.868
  • Benzene:
    Max. 0.02 %
  • Evaporation residue:
    Max. 50 ppm

Specification Test Results

About this item

CAS Number:108-88-3
 :
Formula:C₆H₅CH₃
MDL Number:MFCD00008512
 :92.14 g/mol
Merck Index:13,09607
Boiling Pt:110.6 °C (1013 hPa)
Melting Pt:–95 °C
Density:0.867 g/cm³ (20 °C)
Flash Pt:4 °C
Storage Temperature:Ambient
EINECS:203-625-9
UN:1294
ADR:3,II
REACH:01-2119471310-51
 :GHS02
 :GHS08
 :GHS07