ContactUsLinkComponent
Tubing connectors, Magic box, LaboPlast®
  229-7420
 :  BURKLE
undefined
Tubing connectors, Magic box, LaboPlast®
  229-7420
 :  BURKLE
  • Tubing int.Ø:
    4 - 17 mm
  • Pk:
    1 SET
Magic box contains 68 connectors.
  • Good chemical resistance
  • Good temperature resistance
In this magic box, you can invariably find the appropriate hose connector you are looking for.
 : Includes straight hose connector, T hose connector,Y hose connector, X hose connector, angle hose connector (two of each in sizes 4, 6, 8, 10, 12 and 14 mm), universal hose connector (two of each for hoses from 4 to 17 mm), reducing hose connector (two of each in sizes 4 to 8, 4 to 12, and 8 to 12 mm).