Orders LinkContactUsLinkComponent
Non return valves
Supplier:   BURKLE
Non return valves
Supplier:  BURKLE
Certificates
Non return valves
Non return valves
6 Options Available Below
Product Details & Documents
Translucent PP retaining valves with embossed directional arrow or printed in black. With tapered ends. Various uses.
  • Resistant to compression max. 4 bar
  • Operating temperature up to +40 °C
  • With PP ball valve
Product Family Options
Product Information
  • Tubing size
    Availability
    Price
  • 229-0134
    Non return valve with embossed directional arrow
    Non return valve with embossed directional arrow
    6 - 8 mm ID
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2,90 €
    2,90 €
    1 * 1 ST
       2,90 €
  • BURK8714-1006
    Non return valve with black printed directional arrow
    Non return valve with black printed directional arrow
    6 - 8 mm ID
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3,20 €
    3,20 €
    1 * 1 ST
       3,20 €
  • 229-0135
    Non return valve with embossed directional arrow
    Non return valve with embossed directional arrow
    8 - 10 mm ID
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2,90 €
    2,90 €
    1 * 1 ST
       2,90 €
  • 229-0137
    Non return valve with black printed directional arrow
    Non return valve with black printed directional arrow
    8 - 10 mm ID
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3,20 €
    3,20 €
    1 * 1 ST
       3,20 €
  • 229-0136
    Non return valve with embossed directional arrow
    Non return valve with embossed directional arrow
    10 - 14 mm ID
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3,70 €
    3,70 €
    1 * 1 ST
       3,70 €
  • BURK8714-1010
    Non return valve with black printed directional arrow
    Non return valve with black printed directional arrow
    10 - 14 mm ID
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3,90 €
    3,90 €
    1 * 1 ST
       3,90 €