Toluene ≥99.7% (by GC), CHROMASOLV™ for pesticide residue analysis acc. to FDA, Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:108-88-3
:
Formula:C₆H₅CH₃
MDL Number:MFCD00008512
:92.14 g/mol
Merck Index:13,09607
Boiling Pt:110.6 °C
Melting Pt:–95 °C
Density:0.867 g/cm³ (20 °C)
Flash Pt:7.1 %(V)
Storage Temperature:Ambient
EINECS:203-625-9
UN:1294
ADR:3,II
REACH:05-2114615130-69
:GHS07
:GHS08
:GHS02
Specification Test Results
- Assay (GC):Min. 99.7 %
- Non-volatile matter:Max. 0.0005 %
- Water (Karl Fischer):Max. 0.02 %
- Suitability for residue analysis:Max. 5 ng/l
- Refractive Index:n20/D 1.496 (lit.)
- Explosion Limit:7 %
- Autoignition Temperature:997 °F
- Vapor Pressure:22 mmHg ( 20 °C)
Specification Test Results
Frequently Bought Together