Order Entry
Austria
ContactUsLinkComponent
 
 
 
 
Toluene ≥99.7%, CHROMASOLV™ for analysis of dioxins, furans and PCB, Riedel-de Haën™
 
undefined
 
 
 
Toluene ≥99.7%, CHROMASOLV™ for analysis of dioxins, furans and PCB, Riedel-de Haën™

 

  • Assay (GC):
    Min. 99.7 %
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.02 %
  • Suit.f.GC-MS of dioxines,furanes,PCBs:
    complying
  • Vapor Pressure:
    22 mmHg ( 20 °C)
  • Explosion Limit:
    7 %
  • Vapor Density:
    3.2 (vs air)

 

 

Frequently Bought Together