


Toluene, BAKER, Laboratory Reagent, J.T.Baker®
Artikelnr. 8078.2500
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Assay (by GC):min. 98 %
- Boiling Range:109 - 112 °C
- Residue after Evaporation:max. 0.002 %
- Water (H2O):max. 0.05 %
Specification Test Results
About this item
:
:92.14 g/mol
Merck Index:13,09607
Boiling Pt:110.6 °C (1013 hPa)
Melting Pt:–95 °C
Density:0.867 g/cm³ (20 °C)
Flash Pt:4 °C
Storage Temperature:Ambient
EINECS:203-625-9
UN:1294
ADR:3,II
REACH:05-2114615130-69
:GHS02
:GHS08
:GHS07
CAS Number:108-88-3
Formula:C₆H₅CH₃
MDL Number:MFCD00008512