Hose tubing connectors, straight, cylindrical nozzles, LaboPlast®
Supplier: BURKLE
Certificates
About this item
Cylindrical straight hose connectors for various hose sizes. PP or PVDF, translucent.
- PP, temperature resistant up to +135 °C max.
- PVDF, temperature resistant up to +160 °C max., highly resistant to chemicals
- Sterilisable
PVDF connectors with food industry approval, autoclavable and UV-resistant.
Frequently Bought Together