Stopcock, two way, double oblique bore DURAN®
Lieferant: DWK Life Sciences
Zertifikate
Über diesen Artikel
DURAN®, borosilicate glass 3.3, clear, with glass or PTFE key.
- Very good chemical resistance
- Very good temperature resistance
Two oblique bores.
Frequently Bought Together