Screw caps for T501 sample tubes
Supplier: Simport Scientific
Certificates
About this item
Caps are available either with or without a silicone washer between the cap and the tube to ensure a positive leakproof seal at all temperatures.
- Made of polypropylene
- Available with or without silicone washer
- Long skirt and super fast thread design for easy seal
- Autoclavable
As for tubes, these closures are also made of polypropylene, providing the same coefficient of expansion for both, which further enhances the leakproof qualities of the vials at changing temperatures. The cap features a long skirt and a super-fast thread design that allows it to be removed or sealed with a single turn.
Ordering information: Tubes T501 series sold separately.
Frequently Bought Together