Sample tubes
Supplier: Simport Scientific
Jump to:
About this item
PP, non toxic, translucent.
- Very good chemical resistance
- Smooth and uniform inner surface
- Tubes can be autoclaved (121° C) in upright position with caps loosened
- Round bottom tubes only can be centrifuged up to 17000×g
Designed for the storage and transportation of biological material.
*For size only, cap is included.
16 Options Available Below
Product Details & Documents
PP, non toxic, translucent.
- Very good chemical resistance
- Smooth and uniform inner surface
- Tubes can be autoclaved (121° C) in upright position with caps loosened
- Round bottom tubes only can be centrifuged up to 17000×g
Designed for the storage and transportation of biological material.
*For size only, cap is included.
Documents
Certificates
Search by Lot NumberProduct Family Options
Tubes, not printed
Product Information
- CapacityClosure typeSizeTypeAvailabilityPrice
- 212-9336With external thread, ungraduatedWith external thread, ungraduated1,2 mlWithout cap12,5×43 mmSelf standingLog in for availability1 * 1.000 ST236,00 €
- 212-9338With external thread, ungraduatedWith external thread, ungraduated2 mlWithout cap12,5×48 mmRound bottomLog in for availability1 * 1.000 ST220,00 €
- 212-9337With external thread, ungraduatedWith external thread, ungraduated2 mlWithout cap12,5×49 mmSelf standingLog in for availability1 * 1.000 ST207,00 €
- 212-9339With external thread, ungraduatedWith external thread, ungraduated3 mlWithout cap12,5×72 mmSelf standingLog in for availability1 * 1.000 ST334,00 €
- 212-9341With external thread, ungraduatedWith external thread, ungraduated4 mlWithout cap12,5×75 mmRound bottomLog in for availability1 * 1.000 ST329,00 €
- 212-9340With external thread, ungraduatedWith external thread, ungraduated4 mlWithout cap12,5×76 mmSelf standingLog in for availability1 * 1.000 ST329,00 €
- 212-9343With external thread, ungraduatedWith external thread, ungraduated5 mlWithout cap12,5×92 mmRound bottomLog in for availability1 * 1.000 ST335,00 €
- 212-9342With external thread, ungraduatedWith external thread, ungraduated5 mlWithout cap12,5×93 mmSelf standingLog in for availability1 * 1.000 ST279,00 €
Tubes, graduated, white printed
Product Information
- CapacityClosure typeSizeTypeAvailabilityPrice
- SIMPT501-1ATPRWith external thread, graduatedWith external thread, graduated1,2 mlWithout cap12,5×43 mmSelf standingLog in for availability1 * 1.000 ST374,00 €
- SIMPT501-2TPRWith external thread, graduatedWith external thread, graduated2 mlWithout cap12,5×48 mmRound bottomLog in for availability1 * 1.000 ST374,00 €
- SIMPT501-2ATPRWith external thread, graduatedWith external thread, graduated2 mlWithout cap12,5×49 mmSelf standingLog in for availability1 * 1.000 ST374,00 €
- SIMPT501-3ATPRWith external thread, graduatedWith external thread, graduated3 mlWithout cap12,5×72 mmSelf standingLog in for availability1 * 1.000 ST419,00 €
- SIMPT501-4TPRWith external thread, graduatedWith external thread, graduated4 mlWithout cap12,5×75 mmRound bottomLog in for availability1 * 1.000 ST422,00 €
- SIMPT501-4ATPRWith external thread, graduatedWith external thread, graduated4 mlWithout cap12,5×76 mmSelf standingLog in for availability1 * 1.000 ST422,00 €
- SIMPT501-5TPRWith external thread, graduatedWith external thread, graduated5 mlWithout cap12,5×92 mmRound bottomLog in for availability1 * 1.000 ST422,00 €
- SIMPT501-5ATPRWith external thread, graduatedWith external thread, graduated5 mlWithout cap12,5×93 mmSelf standingLog in for availability1 * 1.000 ST422,00 €
Caps
Product Information
- Catalog #MaterialDescriptionAvailabilityPrice