Orders LinkContactUsLinkComponent
Containers, multi-purpose, with screw caps
Containers, multi-purpose, with screw caps
120 ml, Sterile, Red, individually packed, Carton of 250
Artikelnr. RATI5921183
Anbieter:   RATIOLAB
Containers, multi-purpose, with screw caps
Artikelnr. RATI5921183
Anbieter:  RATIOLAB
Containers, multi-purpose, with screw caps
Containers, multi-purpose, with screw caps
120 ml, Sterile, Red, individually packed, Carton of 250
Preis
72,70 €
Log in for availability
1 * 250 ST
Product Details & Documents
Made of PP with red screw-cap.
  • Multi-purpose
  • Ideal for the clinical routine
  • Good chemical and temperature resistance
Suitable for sampling, transport and storage of solid, pasty and liquid samples.
Specifications
  • Packed:
    individually packed, Carton of 250
  • Type:
    Sterile
  • Capacity:
    120 ml
  • Seq:
    02
  • Cap colour:
    Red