Orders LinkContactUsLinkComponent
Screw caps for internal thread sample tubes
Screw caps for internal thread sample tubes
Supplier:   Simport Scientific
Screw caps for internal thread sample tubes
Supplier:  Simport Scientific
Certificates
Screw caps for internal thread sample tubes
Screw caps for internal thread sample tubes
8 Options Available Below
Product Details & Documents
These polypropylene screw caps are to be used with T500 series internal thread sample tubes.
  • Made of polypropylene
  • Cap with silicone washer for leakproof seal
  • Long skirt and super-fast thread design for easy seal
  • Autoclavable
Caps are available with a silicone washer between the cap and the tube to ensure a positive leakproof seal at all temperatures. The cap features a long skirt and a super-fast thread design allowing it to be removed or screwed on with a single turn.
These silicone washer caps will guarantee a positive leakproof seal at all temperatures.
Ordering information: Tubes T500 series sold separately.
Product Family Options
Product Information
  • Colour
    Availability
    Price
  • 212-9335
    Screw caps with silicone washer
    Screw caps with silicone washer
    Natural
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 246,00 €
    246,00 €
    1 * 1.000 ST
       246,00 €
  • SIMPT500ROS
    Screw caps with silicone washer
    Screw caps with silicone washer
    Red
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 246,00 €
    246,00 €
    1 * 1.000 ST
       246,00 €
  • SIMPT500YOS
    Screw caps with silicone washer
    Screw caps with silicone washer
    Yellow
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 246,00 €
    246,00 €
    1 * 1.000 ST
       246,00 €
  • SIMPT500BOS
    Screw caps with silicone washer
    Screw caps with silicone washer
    Blue
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 246,00 €
    246,00 €
    1 * 1.000 ST
       246,00 €
  • SIMPT500GOS
    Screw caps with silicone washer
    Screw caps with silicone washer
    Green
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 246,00 €
    246,00 €
    1 * 1.000 ST
       246,00 €
  • SIMPT500WOS
    Screw caps with silicone washer
    Screw caps with silicone washer
    White
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 246,00 €
    246,00 €
    1 * 1.000 ST
       246,00 €
  • SIMPT500LOS
    Screw caps with silicone washer
    Screw caps with silicone washer
    Lilac
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 246,00 €
    246,00 €
    1 * 1.000 ST
       246,00 €
  • SIMPT500OOS
    Screw caps with silicone washer
    Screw caps with silicone washer
    Orange
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 246,00 €
    246,00 €
    1 * 1.000 ST
       246,00 €