Order Entry
Austria
ContactUsLinkComponent
Accessories for AQ3070, pH indicator
Accessories for AQ3070, pH indicator
Catalog # ORIOAC3001
Supplier:  Thermo Orion
CAS Number:  
undefined
Accessories for AQ3070, pH indicator
Catalog # ORIOAC3001
Supplier:  Thermo Orion
CAS Number:  

Specifications

  • Pk:
    1
  • Description:
    pH indicator (2×15 ml)
  • Tests per kit:
    30 tests
  • Range:
    5,9 - 8,2 pH

Specifications

About this item

For use with Orion AquaMate 8000 UV-Vis Spectrophotometer, Orion AquaMate 7000 Vis Spectrophotometer, Orion AQUAfast AQ3700 colorimeter, Orion AQUAfast AQ3070