Order Entry
Austria
ContactUsLinkComponent
Vortex mixers, CLASSIC and WIZARD
Vortex mixers, CLASSIC and WIZARD
Supplier:  VELP SCIENTIFIC
Certificates
undefined
Vortex mixers, CLASSIC and WIZARD
Supplier:  VELP SCIENTIFIC

Specifications

  • Catalog No:
  • 444-0746
  • 444-0745
  • Model:
  • WIZARD
  • CLASSIC
  • Shaking motion:
  • Orbital
  • Orbital
  • Orbit:
  • 4,5 mm
  • 4,5 mm
  • Speed range:
  • 0 - 3000 min⁻¹
  • 0 - 3000 min⁻¹
  • Input power:
  • 15 W
  • 15 W
  • W×D×H:
  • 180×220×70 mm
  • 180×220×70 mm
  • Weight:
  • 2,4 kg
  • 2,4 kg

Specifications

Related Information

Frequently Bought Together