ContactUsLinkComponent
Tubing connectors, complete with screw cap, straight, DURAN®
Tubing connectors, complete with screw cap, straight, DURAN®
Catalog # SCOT286221806
Supplier:  DWK Life Sciences
undefined
Tubing connectors, complete with screw cap, straight, DURAN®
Catalog # SCOT286221806
Supplier:  DWK Life Sciences
Specifications
  • Thread:
    GL 14
  • Tubing int.Ø:
    8 mm
  • Pk:
    10
Specifications
About this item
Screw cap PTB, red, with white PP straight hose connection.
  • Very good chemical resistance
  • Very good temperature resistance
With silicone seal (VMQ).