Orders LinkContactUsLinkComponent
Hose clips, worm-threaded
Hose clips, worm-threaded
Supplier:   BURKLE
Hose clips, worm-threaded
Supplier:  BURKLE
Certificates
Hose clips, worm-threaded
Hose clips, worm-threaded
6 Options Available Below
Product Details & Documents
Hose clip sleeve made of stainless steel 1,4016, screw zinc coated, with screw thread.
Product Family Options
Product Information
  • Spanning width
    Availability
    Price
  • 8 - 12 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 14,00 €
    14,00 €
    1 * 10 ST
       14,00 €
  • 10 - 16 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 14,10 €
    14,10 €
    1 * 10 ST
       14,10 €
  • 12 - 20 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 16,30 €
    16,30 €
    1 * 10 ST
       16,30 €
  • 16 - 27 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 16,60 €
    16,60 €
    1 * 10 ST
       16,60 €
  • 20 - 32 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 16,80 €
    16,80 €
    1 * 10 ST
       16,80 €
  • 25 - 40 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 17,20 €
    17,20 €
    1 * 10 ST
       17,20 €