Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Stopcocks
Stopcocks, NO-DRIP
Stopcocks, NO-DRIP
216-0235
:
BOCHEM
Stopcocks, NO-DRIP
216-0235
:
BOCHEM
Description:
"Zero drip" tap made of 18/10 stainless steel
Pk:
1
This stopcock are already mounted, PTFE seal with max. 260 °C.