Orders LinkContactUsLinkComponent
Wire baskets, rectangular, with lid
Wire baskets, rectangular, with lid
Wire baskets, rectangular, with lid
Wire baskets, rectangular, with lid
These fine mesh baskets, made of stainless steel, electropolished, are designed to keep small parts and accessories.
  • With lid
  • With base frame
  • Mesh size: 1,6×1,6 mm
Colour: Silver.
Product Family Options
  • W×D×H
  • 80×40×30 mm
    1 * 1 ST
    1 * 1 ST - 231,00 €
    231,00 €
    1 * 1 ST
       231,00 €
  • 80×80×40 mm
    1 * 1 ST
    1 * 1 ST - 237,00 €
    237,00 €
    1 * 1 ST
       237,00 €
  • 210×150×42 mm
    1 * 1 ST
    1 * 1 ST - 547,00 €
    547,00 €
    1 * 1 ST
       547,00 €
  • 200×100×50 mm
    1 * 1 ST
    1 * 1 ST - 392,00 €
    392,00 €
    1 * 1 ST
       392,00 €
  • 220×140×50 mm
    1 * 1 ST
    1 * 1 ST - 384,00 €
    384,00 €
    1 * 1 ST
       384,00 €
  • 274×174×36 mm
    1 * 1 ST
    1 * 1 ST - 402,00 €
    402,00 €
    1 * 1 ST
       402,00 €
  • 310×210×40 mm
    1 * 1 ST
    1 * 1 ST - 505,00 €
    505,00 €
    1 * 1 ST
       505,00 €