ContactUsLinkComponent
Stand for Imhoff funnels
Stand for Imhoff funnels
Catalog # BURK7118-2000
Supplier:  BURKLE
undefined
Stand for Imhoff funnels
Catalog # BURK7118-2000
Supplier:  BURKLE
Specifications
  • Description:
    for two Imhoff funnels
  • Pk:
    1
Specifications
About this item
PP, chrome-plated steel. The stand for sedimentation funnels is used to keep, hold or store Imhoff funnels safely and reliably during sedimentation.
  • Adjustable height up to 450 mm
  • For funnels with Ø 90 mm or more
Two funnel holders ensure that Imhoff funnels will be held safely and securely and can be placed in an absolutely vertical position. Thanks to an innovative magnet concept no clamping screws are necessary. The height of the funnel holder can be adjusted easily and flexibly with one hand. With the corresponding accessory the holders for Imhoff funnels can also be replaced by holders for all-purpose funnels.