

Benzylamine for synthesis, Sigma-Aldrich®
: Merck
:100-46-9
:
:(C₆H₅)CH₂NH₂
:MFCD00008106
:107.16 g/mol
:13,01127
:185 °C (1013 hPa)
:10 °C
:0.98 g/cm³ (20 °C)
:202-854-1
:2735
:8,II
:M-Clarity™ quality level = MQ200
:65 °C
:GHS05
:GHS07
:Ambient
- Solubility:(20 °C) soluble
- Melting Point:-42.8 °C
- Molar Mass:107.15 g/mol
- Boiling Point:185 °C
- Vapor Pressure:1.2 hPa (20 °C)
- Flash Point:72 °C
- Refractive Index:1.5438 (20 °C, 589 nm)
- Explosion Limit:0.7 - 8.2 %(V)
- Density:0.98 g/cm3 (20 °C)
- pH:11.4 (100 g/l, H2O, 20 °C)
- Ignition Point:405 °C
- Synonyms:Phenylmethylamine
- Hill Formula:C7H9N
Frequently Bought Together