Order Entry
Austria
Orders LinkContactUsLinkComponent
2-Iodotoluene

2-Iodotoluene

Glass bottle for solids, 100 g
Catalog # MOLE46951044-100G
Irritant
Marketsource
Supplier:   Molekula
2-Iodotoluene
Catalog # MOLE46951044-100G
Supplier:  Molekula
Marketsource
CAS Number:  615-37-2
2-Iodotoluene

Glass bottle for solids, 100 g

CAS Number:  615-37-2
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  1-Iodo-2-methylbenzene
  • CAS Number:615-37-2
  • Formula:C₇H₇I
  • MDL Number:MFCD00001042
  • Molecular Weight:218.04 g/mol
  • Boiling Pt:211 °C (1013 hPa)
  • Density:1,71 g/cm³ (20 °C)
  • Flash Pt:90 °C
  • Storage Temperature:Ambient
  • EINECS:210-422-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications