Tubing, silicone, Versitec®
Supplier: Saint Gobain Life Sciences
Certificates
About this item
Silicone, peroxide cured, translucent.
- Water-repellent
- Anti-adhesive
- Exceptional resistance to UV rays, oxygen and ozone
- Ppermeable.
Versitec tubing is a high quality industrial silicone used in food industry, household electrical appliances, electrical industry and in drink dispensers. The technical performance of the Versitec® tubing, together with its effectiveness make it the ideal means of transferring high purity or high-tech fluids.
Temperature resistance: -60 to +200 °C
Hardness (Shore durometer A): 40
Density: 1,15
Certifications: Compliant with food standards French, German (BfR) and American (FDA)
Frequently Bought Together