ContactUsLinkComponent
Clips assortment for spherical ground joints, POM, KECK™, DURAN®
Clips assortment for spherical ground joints, POM, KECK™, DURAN®
Catalog # SCOT290320007
Supplier:  DWK Life Sciences
Clips assortment for spherical ground joints, POM, KECK™, DURAN®
Catalog # SCOT290320007
Supplier:  DWK Life Sciences
Specifications
  • For NS:
    13 / 19 / 29 / 35
  • Colour:
    Assorted
  • Pk:
    1
Specifications
About this item
Polyoxymethylene, coloured.
Delivery: The set includes the following clips: 2 × KS 13, 2 × KS 19, 1 × KS 29, 1 × KS 35.