ContactUsLinkComponent
Clips assortment for conical joints, POM, KECK™, DURAN®
Clips assortment for conical joints, POM, KECK™, DURAN®
Catalog # SCOT290310006
Supplier:  DWK Life Sciences
Clips assortment for conical joints, POM, KECK™, DURAN®
Catalog # SCOT290310006
Supplier:  DWK Life Sciences
Specifications
  • For NS:
    10 / 14 / 19 / 24 / 29 / 34 / 45
  • Colour:
    Assorted
  • Pk:
    1
Specifications
About this item
Polyoxymethylene, coloured.
Delivery: The set includes the following clips: 2 × KC 14, 2 × KC 19, 1 × KC 29, 1 × KC 10, 1 × KC 24, 1 × KC 34, 1 × KC 45.