Sponge, VertiKlean®
Über diesen Artikel
The VertiKlean Sponge is a disposable laboratory sponge. It consists of Polynit wiping material that has been laminated to a clean polyester foam. The sponge is laser cut to completely seal the edges. It is designed for general cleaning and wiping but can also be used in laboratories.
- High sorbency polyester foam with the abrasion resistance of Polynit fabric
- Autoclavable
2 Optionen unten verfügbar
Product Details & Documents
The VertiKlean Sponge is a disposable laboratory sponge. It consists of Polynit wiping material that has been laminated to a clean polyester foam. The sponge is laser cut to completely seal the edges. It is designed for general cleaning and wiping but can also be used in laboratories.
- High sorbency polyester foam with the abrasion resistance of Polynit fabric
- Autoclavable
Product Family Options
Produktinformationen