ContactUsLinkComponent
Dressing pads for catheter fixation, BD Veca-C™, BD Vecafix™
Dressing pads for catheter fixation, BD Veca-C™, BD Vecafix™
Catalog # BDAM392020UK
Supplier:  BD Medical
Dressing pads for catheter fixation, BD Veca-C™, BD Vecafix™
Catalog # BDAM392020UK
Supplier:  BD Medical
Specifications
  • Description:
    Dressing pad, BD Veca-C™, 75×60 mm, with transparent window, UK plug
  • Seq:
    02
  • Pk:
    600
Specifications
About this item
BD Veca-C™ and BD Vecafi x™ are sterile IV dressings for optimum fixation of ported and non-ported IV catheters.
  • Single use
  • Sterile
  • Latex free
It protects the insertion site and allows the site to breathe. The wound pad absorbs exudate at the
insertion site, keeping the site dry. Sterilisation method is ethylene oxide. BD Veca-C™ has a transparent window of polyurethane that allows for visualization of phlebitis along the catheter track.