

Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for preparative HPLC
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:141-78-6
:
Formula:CH₃COOC₂H₅
MDL Number:MFCD00009171
:88.11 g/mol
Merck Index:13,03792
Boiling Pt:77.1 °C (1013 hPa)
Melting Pt:–83 °C
Density:0.902 g/cm³ (20 °C)
EINECS:205-500-4
UN:1173
ADR:3,II
:Filtered through a 0.2 µm filter, packaged under nitrogen.
Flash Pt:–4 °C
REACH:01-2119475103-46
:GHS02
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
- Assay (calculated on anhydrous):Min. 99.8 %
- Acidity:Max. 0.0005 meq/g
- Evaporation residue:Max. 0.0005 %
- Water:Max. 0.02 %
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC84532.460 spec_VWRC84532.460 spec_VWRC84532.550 spec_VWRC84532.950
Product specification
VWR® for chromatography columns, consumables and reagents
Product Literature
Frequently Bought Together