PCR racks for tubes, strips and plates, PCRack™
Supplier: Simport Scientific
Certificates
About this item
PCRack™ made of PP. Suitable for PCR individual tubes, 96 well plate and strips of 8 or 12 tubes.
- Space saving
- Stackable
It can be horizontally attached to each other in order to build-up any configuration. With the cover on, they are easily stackable one on top of another.
Frequently Bought Together