Triple clamps
Supplier: BOCHEM
Certificates
About this item
Holder for Schott laboratory flasks for the alternating removal of vessel and lid.
This special quick-release fastener ensures that the two laboratory flat flanges of reaction tube and flat flange lid are connected and, in addition, that the reaction tube and/or flat flange lid are held individually (optimal flange angle 45°).
Frequently Bought Together