Vacuum hand pumps, VacuMan
Supplier: BURKLE
Certificates
About this item
The vacuum hand pump can be operated with one hand to create a vacuum or pressure, independent of power supply.
- Maintenance-free, self-lubricating
- Corrosion resistant
- With or without gauges
- Available in PVC or aluminium for heavy duty
Connection for hoses with 6,4 mm inner diameter.
Accessories information: Sampling set with suction hose, sample container and adapter set for hoses from 5 to 14 mm available as accessory
Frequently Bought Together