Order Entry
Austria
ContactUsLinkComponent
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for preparative HPLC
Flammable
Irritant
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for preparative HPLC
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates
undefined
Flammable
Irritant
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for preparative HPLC
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default

Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99.9 %
  • Acidity:
    Max. 0.0005 meq/g
  • Evaporation residue:
    Max. 5 ppm
  • Water:
    Max. 0.02 %

Specification Test Results

Frequently Bought Together