PCR plates, 96-well, raised rim, Amplate™
Supplier: Simport Scientific
Certificates
About this item
Amplate™ raised rim, thin walled PCR plates offer the rigidity needed for automation and are perfectly suited for high performance thermal cycling. Each well makes intimate contact with the heating block while quick and consistent heat transfer is ensured by a uniform wall thickness.
- PP
- Using polypropylene with low adhesion surface properties ensures samples are easily recovered
- An alphanumeric grid helps sample identification
All sealing methods can be used for oil-free operation: Domed and flat cap strips; SecureSeal™ Thermal Sealing Film; SecureSeal™ Aluminium Sealing Foil and Amplate™ Mat. To facilitate orientation, one corner of the plate is cut away.
Delivery: Packed in tamperproof resealable bags of ten plates.
Frequently Bought Together