Order Entry
Austria
ContactUsLinkComponent
Multi-tube vortex mixer, V-32
Multi-tube vortex mixer, V-32
Catalog # 444-0939
Supplier:  GRANT INSTRUMENTS
undefined
Multi-tube vortex mixer, V-32
Catalog # 444-0939
Supplier:  GRANT INSTRUMENTS

Specifications

  • Max. capacity:
    Up to 32 tubes: 16×1,5 ml + 8×0,5 ml + 8×0,2 ml
  • Orbit:
    2 mm
  • Description:
    Multi-tube vortex mixer V-32
  • Model:
    V-32
  • Speed:
    500 - 3000 min⁻¹
  • Weight:
    1,5 kg
  • W×D×H:
    120×180×100 mm
  • Pk:
    1

Specifications

About this item

Versatile multi-tube vortex mixer for vigorous re-suspension of cells or chemical pellets in a maximum of 32 tubes (16×1,5 ml, 8×0,5 ml and 8×0,2 ml), with the facility to mix individual tubes up to 15 ml. Typical applications include vigorous stirring and re-suspension of bacterial and yeast cells and pellets, metabolite and enzyme extraction and various DNA operations such as deproteinisation of DNA/protein complexes and purification of low molecular weight DNA/RNA fragments.

  • Easy to use - continuous or touch operation and set speed knob from 500 to 3000 rpm
  • Orbit: 2 mm
  • Compact rugged design plus powerful motor gives quiet and consistent performance
  • Rubber suction pads prevent the unit from ‘walking’ and also absorb vibration
Delivery: Supplied with universal 32-place platform and flat platform for single tube mixing.