Orders LinkContactUsLinkComponent
Removable needles
Removable needles
New Product
Supplier:   HAMILTON BONADUZ
Removable needles
Supplier:  HAMILTON BONADUZ
New Product
Certificates
Removable needles
Removable needles
83 Options Available Below
Product Details & Documents
The benefit of a replaceable needle for the occasional bend or clog and the minimal dead volume of a cemented needle. The unique removable needle design installs the needle precisely at the zero line. The result is a zero dead volume connection that simplifies priming.
Custom lengths and point styles are available. You can order your custom made needle according to your specification.
* Large hub
** Kel-F hub
Product Family Options
RN fused silica needles for 5 - 10 µl syringes
Product Information
  • Gauge
    Length
    Ext.Ø
    Ø int.
    Type of tip
    Availability
    Price
  • FS 0,17
    100 mm
    0,17 mm
    0,08 mm
    3
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 114,00 €
    114,00 €
    1 * 3 ST
       114,00 €
  • FS 0,23
    100 mm
    0,23 mm
    0,08 mm
    3
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 133,00 €
    133,00 €
    1 * 3 ST
       133,00 €
RN PEEK needles for 250 µl - 10 ml syringes
Product Information
  • Gauge
    Length
    Ext.Ø
    Ø int.
    Type of tip
    Availability
    Price
  • 34 *
    51 mm
    1,52 mm
    0,76 mm
    3
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 256,00 €
    256,00 €
    1 * 6 ST
       256,00 €
  • 34 **
    51 mm
    1,52 mm
    0,76 mm
    3
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 288,00 €
    288,00 €
    1 * 6 ST
       288,00 €
RN needles for 2,5 - 100 µl syringes
Product Information
  • Gauge
    Length
    Ext.Ø
    Ø int.
    Type of tip
    Availability
    Price
  • 26s
    51 mm
    0,47 mm
    0,13 mm
    2
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 63,90 €
    63,90 €
    1 * 6 ST
       63,90 €
  • 26
    51 mm
    0,46 mm
    0,26 mm
    2
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 63,90 €
    63,90 €
    1 * 6 ST
       63,90 €
  • 22s
    51 mm
    0,72 mm
    0,15 mm
    2
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 66,00 €
    66,00 €
    1 * 6 ST
       66,00 €
  • 22s
    51 mm
    0,72 mm
    0,15 mm
    2
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 95,80 €
    95,80 €
    1 * 6 ST
       95,80 €
  • 22
    51 mm
    0,72 mm
    0,41 mm
    2
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 64,90 €
    64,90 €
    1 * 6 ST
       64,90 €
  • 33
    38,1 mm
    0,21 mm
    0,11 mm
    3
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 203,00 €
    203,00 €
    1 * 6 ST
       203,00 €
  • 32
    51 mm
    0,24 mm
    0,11 mm
    3
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 198,00 €
    198,00 €
    1 * 6 ST
       198,00 €
  • 31
    51 mm
    0,31 mm
    0,13 mm
    3
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 201,00 €
    201,00 €
    1 * 6 ST
       201,00 €
  • 30
    51 mm
    0,31 mm
    0,16 mm
    3
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 198,00 €
    198,00 €
    1 * 6 ST
       198,00 €
  • 28
    51 mm
    0,36 mm
    0,18 mm
    3
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 198,00 €
    198,00 €
    1 * 6 ST
       198,00 €
  • 27
    51 mm
    0,41 mm
    0,21 mm
    3
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 285,00 €
    285,00 €
    1 * 6 ST
       285,00 €
  • 26s
    51 mm
    0,47 mm
    0,13 mm
    3
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 78,30 €
    78,30 €
    1 * 6 ST
       78,30 €
  • 26
    51 mm
    0,46 mm
    0,26 mm
    3
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 78,30 €
    78,30 €
    1 * 6 ST
       78,30 €
  • 22s
    51 mm
    0,72 mm
    0,15 mm
    3
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 76,30 €
    76,30 €
    1 * 6 ST
       76,30 €
  • 22s
    19 mm
    0,72 mm
    0,15 mm
    3
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 81,40 €
    81,40 €
    1 * 6 ST
       81,40 €
  • 22
    51 mm
    0,72 mm
    0,41 mm
    3
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 78,30 €
    78,30 €
    1 * 6 ST
       78,30 €
  • 26s
    51 mm
    0,47 mm
    0,13 mm
    5
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 234,00 €
    234,00 €
    1 * 6 ST
       234,00 €
  • 26
    51 mm
    0,46 mm
    0,26 mm
    5
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 234,00 €
    234,00 €
    1 * 6 ST
       234,00 €
  • 22s
    51 mm
    0,72 mm
    0,15 mm
    5
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 234,00 €
    234,00 €
    1 * 6 ST
       234,00 €
  • 22
    51 mm
    0,72 mm
    0,41 mm
    5
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 234,00 €
    234,00 €
    1 * 6 ST
       234,00 €
RN needles for 250 µl - 10 ml syringes
Product Information
  • Gauge
    Length
    Ext.Ø
    Ø int.
    Type of tip
    Availability
    Price
  • 26s
    51 mm
    0,47 mm
    0,13 mm
    2
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 79,40 €
    79,40 €
    1 * 6 ST
       79,40 €
  • 26
    51 mm
    0,46 mm
    0,26 mm
    2
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 79,40 €
    79,40 €
    1 * 6 ST
       79,40 €
  • 22s
    51 mm
    0,72 mm
    0,15 mm
    2
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 79,40 €
    79,40 €
    1 * 6 ST
       79,40 €
  • 22
    51 mm
    0,72 mm
    0,41 mm
    2
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 79,40 €
    79,40 €
    1 * 6 ST
       79,40 €
  • 26s
    51 mm
    0,47 mm
    0,13 mm
    3
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 79,40 €
    79,40 €
    1 * 6 ST
       79,40 €
  • 26
    51 mm
    0,46 mm
    0,26 mm
    3
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 78,30 €
    78,30 €
    1 * 6 ST
       78,30 €
  • 22s
    51 mm
    0,72 mm
    0,15 mm
    3
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 62,90 €
    62,90 €
    1 * 6 ST
       62,90 €
  • 22
    51 mm
    0,72 mm
    0,41 mm
    3
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 60,80 €
    60,80 €
    1 * 6 ST
       60,80 €
  • 22
    19 mm
    0,72 mm
    0,41 mm
    3
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 72,10 €
    72,10 €
    1 * 6 ST
       72,10 €
  • 26s
    51 mm
    0,47 mm
    0,13 mm
    5
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 234,00 €
    234,00 €
    1 * 6 ST
       234,00 €
  • 26
    51 mm
    0,46 mm
    0,26 mm
    5
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 234,00 €
    234,00 €
    1 * 6 ST
       234,00 €
  • 22s
    51 mm
    0,72 mm
    0,15 mm
    5
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 234,00 €
    234,00 €
    1 * 6 ST
       234,00 €
  • 22
    51 mm
    0,72 mm
    0,41 mm
    5
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 230,00 €
    230,00 €
    1 * 6 ST
       230,00 €
Custom made needles for 2,5 - 100 µl syringes
Product Information
  • Gauge
    Length
    Ext.Ø
    Ø int.
    Type of tip
    Availability
    Price
  • 34
    0,16 mm
    0,06 mm
    2, 3 or 4
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 353,00 €
    353,00 €
    1 * 6 ST
       353,00 €
  • 34
    9,5 mm
    0,16 mm
    0,05 mm
    4
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 370,00 €
    370,00 €
    1 * 6 ST
       370,00 €
    Specifications:
    New Product
    New Product
  • 33
    0,21 mm
    0,11 mm
    2, 3 or 4
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 394,00 €
    394,00 €
    1 * 6 ST
       394,00 €
  • 32
    0,24 mm
    0,11 mm
    2, 3 or 4
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 386,00 €
    386,00 €
    1 * 6 ST
       386,00 €
  • 31
    0,31 mm
    0,13 mm
    2, 3 or 4
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 393,00 €
    393,00 €
    1 * 6 ST
       393,00 €
  • 30
    0,31 mm
    0,16 mm
    2, 3 or 4
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 386,00 €
    386,00 €
    1 * 6 ST
       386,00 €
  • 29
    0,33 mm
    0,18 mm
    2, 3 or 4
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 386,00 €
    386,00 €
    1 * 6 ST
       386,00 €
  • 28
    0,36 mm
    0,18 mm
    2, 3 or 4
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 386,00 €
    386,00 €
    1 * 6 ST
       386,00 €
  • 27
    0,41 mm
    0,21 mm
    2, 3 or 4
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 462,00 €
    462,00 €
    1 * 6 ST
       462,00 €
  • 26
    0,46 mm
    0,26 mm
    5 or AS
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 430,00 €
    430,00 €
    1 * 6 ST
       430,00 €
  • 26
    0,46 mm
    0,26 mm
    2, 3 or 4
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 193,00 €
    193,00 €
    1 * 6 ST
       193,00 €
  • 26s
    0,47 mm
    0,13 mm
    5 or AS
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 421,00 €
    421,00 €
    1 * 6 ST
       421,00 €
  • 26s
    0,47 mm
    0,13 mm
    2, 3 or 4
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 193,00 €
    193,00 €
    1 * 6 ST
       193,00 €
  • 25
    0,51 mm
    0,26 mm
    2, 3 or 4
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 193,00 €
    193,00 €
    1 * 6 ST
       193,00 €
  • 25s
    0,52 mm
    0,15 mm
    2, 3 or 4
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 197,00 €
    197,00 €
    1 * 6 ST
       197,00 €
  • 24
    0,57 mm
    0,31 mm
    2, 3 or 4
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 193,00 €
    193,00 €
    1 * 1 ST
       193,00 €
  • 23
    0,64 mm
    0,34 mm
    2, 3 or 4
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 210,00 €
    210,00 €
    1 * 6 ST
       210,00 €
  • 23s
    0,64 mm
    0,12 mm
    2, 3 or 4
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 197,00 €
    197,00 €
    1 * 6 ST
       197,00 €
  • 22s
    0,72 mm
    0,15 mm
    5 or AS
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 415,00 €
    415,00 €
    1 * 6 ST
       415,00 €
  • 22s
    0,72 mm
    0,15 mm
    2, 3 or 4
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 193,00 €
    193,00 €
    1 * 6 ST
       193,00 €
  • 22
    0,72 mm
    0,41 mm
    5 or AS
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 415,00 €
    415,00 €
    1 * 6 ST
       415,00 €
  • 22
    0,72 mm
    0,41 mm
    2, 3 or 4
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 193,00 €
    193,00 €
    1 * 6 ST
       193,00 €
  • 21
    0,82 mm
    0,51 mm
    2, 3 or 4
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 193,00 €
    193,00 €
    1 * 6 ST
       193,00 €
  • 21
    0,82 mm
    0,51 mm
    5 or AS
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 421,00 €
    421,00 €
    1 * 6 ST
       421,00 €
  • 20
    0,91 mm
    0,60 mm
    2, 3 or 4
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 197,00 €
    197,00 €
    1 * 6 ST
       197,00 €
  • 19
    1,07 mm
    0,69 mm
    2, 3 or 4
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 210,00 €
    210,00 €
    1 * 6 ST
       210,00 €
  • 18
    1,27 mm
    0,84 mm
    2, 3 or 4
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 193,00 €
    193,00 €
    1 * 6 ST
       193,00 €
Custom made needles for 250 µl - 10 ml syringes
Product Information
  • Gauge
    Length
    Ext.Ø
    Ø int.
    Type of tip
    Availability
    Price
  • 26
    0,46 mm
    0,26 mm
    5 or AS
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 421,00 €
    421,00 €
    1 * 6 ST
       421,00 €
  • 26
    0,46 mm
    0,26 mm
    2, 3 or 4
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 200,00 €
    200,00 €
    1 * 6 ST
       200,00 €
  • 26s
    0,47 mm
    0,13 mm
    5 or AS
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 421,00 €
    421,00 €
    1 * 6 ST
       421,00 €
  • 25
    0,51 mm
    0,26 mm
    2, 3 or 4
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 193,00 €
    193,00 €
    1 * 6 ST
       193,00 €
  • 26s
    0,47 mm
    0,13 mm
    2, 3 or 4
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 193,00 €
    193,00 €
    1 * 6 ST
       193,00 €
  • 25s
    0,52 mm
    0,15 mm
    2, 3 or 4
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 196,00 €
    196,00 €
    1 * 6 ST
       196,00 €
  • 24
    0,57 mm
    0,31 mm
    2, 3 or 4
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 193,00 €
    193,00 €
    1 * 6 ST
       193,00 €
  • 23
    0,64 mm
    0,34 mm
    2, 3 or 4
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 193,00 €
    193,00 €
    1 * 6 ST
       193,00 €
  • 23s
    0,64 mm
    0,12 mm
    2, 3 or 4
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 193,00 €
    193,00 €
    1 * 6 ST
       193,00 €
  • 22
    0,72 mm
    0,41 mm
    5 or AS
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 421,00 €
    421,00 €
    1 * 6 ST
       421,00 €
  • 22s
    0,72 mm
    0,15 mm
    5 or AS
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 421,00 €
    421,00 €
    1 * 6 ST
       421,00 €
  • 22
    0,72 mm
    0,41 mm
    2, 3 or 4
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 193,00 €
    193,00 €
    1 * 6 ST
       193,00 €
  • 22s
    0,72 mm
    0,15 mm
    2, 3 or 4
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 193,00 €
    193,00 €
    1 * 6 ST
       193,00 €
  • 21
    0,82 mm
    0,51 mm
    2, 3 or 4
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 193,00 €
    193,00 €
    1 * 6 ST
       193,00 €
  • 20
    0,91 mm
    0,60 mm
    2, 3 or 4
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 193,00 €
    193,00 €
    1 * 6 ST
       193,00 €
Small hub RN CTC needles
Product Information
  • Gauge
    Length
    Ext.Ø
    Ø int.
    Type of tip
    Availability
    Price
  • 22s
    51 mm
    0,72 mm
    0,15 mm
    3
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 37,10 €
    37,10 €
    1 * 3 ST
       37,10 €
  • 22
    51 mm
    0,72 mm
    0,41 mm
    3
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 42,30 €
    42,30 €
    1 * 3 ST
       42,30 €
  • 23s
    43 mm
    0,64 mm
    0,12 mm
    AS
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 114,00 €
    114,00 €
    1 * 6 ST
       114,00 €
  • 26s
    43 mm
    0,47 mm
    0,13 mm
    AS
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 114,00 €
    114,00 €
    1 * 6 ST
       114,00 €