Orders LinkContactUsLinkComponent
Scoops
 :   BURKLE
Scoops
 :  BURKLE
Scoops
Scoops
Sterilisable scoops made of stainless steel AISI 304 (1.4301).
  • Robust
  • Autoclavable up to 121 °C for max. 20 min.
  • Suitable for food contact
Product Family Options
  • Capacity
    Ø
  • 120 ml
    50 mm
    1 * 1 ST
    1 * 1 ST - 28,70 €
    28,70 €
    1 * 1 ST
       28,70 €
  • 450 ml
    75 mm
    1 * 1 ST
    1 * 1 ST - 32,40 €
    32,40 €
    1 * 1 ST
       32,40 €
  • 1000 ml
    100 mm
    1 * 1 ST
    1 * 1 ST - 38,50 €
    38,50 €
    1 * 1 ST
       38,50 €
  • 1500 ml
    120 mm
    1 * 1 ST
    1 * 1 ST - 44,10 €
    44,10 €
    1 * 1 ST
       44,10 €
  • 2500 ml
    150 mm
    1 * 1 ST
    1 * 1 ST - 57,90 €
    57,90 €
    1 * 1 ST
       57,90 €