Burners, Teclu
Supplier: BOCHEM
Certificates
About this item
These burners have air regulation and reach a maximum temperature of 1300 °C.
Certifications: DIN 30665
Frequently Bought Together
Gas cartridges, Campingaz®
Supplier: CAMPING GAZ
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR®, Mini vortex mixer
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Volumetric flasks, class A, blue markings, BLAUBRAND®
Supplier: Brand
Cartridge 220G for cartridge burner
Supplier: USBECK Laborgeräte
Eppendorf® Pipette Holder System
Supplier: EPPENDORF
VWR®, Digital Timer, Four Channels, with Clock
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Vortex mixers, Vortex-Genie®
Supplier: SCIENTIFIC INDUSTRIES ELMIS
Dip slides for surfaces and in liquids, Envirocheck®
Supplier: Merck
VWR®, Beakers, Low Form
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Laboratory Bottles, Narrow Neck, with Screw Cap
Supplier: VWR Collection
Vacuum pump, type N 96, LABOPORT®
Supplier: KNF