Samplers, EasySampler
Supplier: BURKLE
Jump to:
About this item
With EasySampler, it is possible to take multiple samples from bulk goods, quickly and easily.
- Emptying greatly simplified via the tip
- No need to disassemble the sampler (in order to obtain a mixed sample)
- Bulk material must be free-flowing or self-flowing
2 Options Available Below
Product Details & Documents
With EasySampler, it is possible to take multiple samples from bulk goods, quickly and easily.
- Emptying greatly simplified via the tip
- No need to disassemble the sampler (in order to obtain a mixed sample)
- Bulk material must be free-flowing or self-flowing
Product Family Options
Product Information
- MaterialLengthØVolumeAvailabilityPrice
- 231-0581EasySampler, stainless steel V4AEasySampler, stainless steel V4AStainless steel V4A100 cm32 mm60 mlLog in for availability1 * 1 ST739,00 €
- 300-0074EasySampler, stainless steel V4AEasySampler, stainless steel V4AStainless steel V4A105 cm32 mm200 mlLog in for availability1 * 1 ST739,00 €