Orders LinkContactUsLinkComponent
Sample lance
Sample lance
Supplier:   BURKLE
Sample lance
Supplier:  BURKLE
Certificates
Sample lance
2 Options Available Below
Product Details & Documents
Samplers for viscous, paste-like and semi-liquid substances.
  • Optimised opening mechanism (can be completely disassembled)
  • Easy to use and to clean
  • Different layers remain intact particularly with paste-like substances
Designed for use in pharmaceutical applications.
Certifications: DIN 51750.3 and 53242.3
Product Family Options
Product Information
  • Material
    Length
    Ø
    Availability
    Price
  • 300-0076
    Valve lance
    Valve lance
    Stainless steel V4A
    550 mm
    32 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 650,00 €
    1 650,00 €
    1 * 1 ST
       1 650,00 €
  • 300-0077
    Valve lance
    Valve lance
    Stainless steel V4A
    800 mm
    32 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 060,00 €
    2 060,00 €
    1 * 1 ST
       2 060,00 €