0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Closures
Bottle Closures
Snap caps, Corning® Gosselin™
Snap caps, Corning® Gosselin™
GOSSC400-08
:
Corning
Low Manufacturing Impact
Snap caps, Corning® Gosselin™
GOSSC400-08
:
Corning
Low Manufacturing Impact
Cap Size:
85 mm
Colour:
Natural
Material:
LDPE
Seq:
2
Environmentally Preferable:
Low Manufacturing Impact
Pk:
1.232
Environmental certification:
RECs, ISO 14001 - Borre