Orders LinkContactUsLinkComponent
Sterilisation Boxes for Pipettes
Sterilisation Boxes for Pipettes
Supplier:   BOCHEM
Sterilisation Boxes for Pipettes
Supplier:  BOCHEM
Certificates
Sterilisation Boxes for Pipettes
Sterilisation Boxes for Pipettes
6 Options Available Below
Product Details & Documents
Stainless steel.
  • Rotating the lid opens or closes the box
  • Sterilisable up to 205 °C
  • Sturdy
Round sterilising pipette cans with silicone covered bottom and lid.
Product Family Options
Product Information
  • Ø×L
    Availability
    Price
  • BOCH8980
    Round Base, for Pipettes up to 382 mm
    Round Base, for Pipettes up to 382 mm
    50×390 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 106,00 €
    106,00 €
    1 * 1 ST
       106,00 €
  • BOCH8981
    Round Base, for Pipettes up to 417 mm
    Round Base, for Pipettes up to 417 mm
    50×425 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 107,00 €
    107,00 €
    1 * 1 ST
       107,00 €
  • 612-1622
    Round Base, for Pipettes up to 372 mm
    Round Base, for Pipettes up to 372 mm
    65×380 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 109,00 €
    109,00 €
    1 * 1 ST
       109,00 €
  • BOCH8983
    Round Base, for Pipettes up to 442 mm
    Round Base, for Pipettes up to 442 mm
    65×450 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 113,00 €
    113,00 €
    1 * 1 ST
       113,00 €
  • BOCH8984
    Round Base, for Pipettes up to 372 mm
    Round Base, for Pipettes up to 372 mm
    80×380 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 116,00 €
    116,00 €
    1 * 1 ST
       116,00 €
  • BOCH8985
    Round Base, for Pipettes up to 492 mm
    Round Base, for Pipettes up to 492 mm
    80×500 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 124,00 €
    124,00 €
    1 * 1 ST
       124,00 €