Deep well plates, 384 square well, Bioblock™
Supplier: Simport Scientific
Certificates
About this item
PP. This 384-well plate is ideal for compound storage and handling of biological samples.
- Round well bottom facilitates aspiration
- Suitable to be used with DMSO and biological buffers
Delivery information: Packed in a bag of 4 plates.
Frequently Bought Together