Order Entry
Austria
Orders LinkContactUsLinkComponent
Scissors for cutting tubes

Scissors for cutting tubes

125 mm
Catalog # BOCH4252
Supplier:   BOCHEM
Scissors for cutting tubes
Catalog # BOCH4252
Supplier:  BOCHEM
Scissors for cutting tubes
Scissors for cutting tubes

125 mm

Price
27,70 €
Log in for availability
1 * 1 ST
Product Details & Documents

Scissors with stainless steel blades and red/black PP handle. Cuts tubes until 10 mm.

Specifications
  • Length:
    125 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.