Order Entry
Austria
ContactUsLinkComponent
Phenol, Calbiochem®
Toxic
Corrosive
Health
Phenol, Calbiochem®
Catalog # 516724-500
CAS Number:  
Toxic
Corrosive
Health
Phenol, Calbiochem®
Catalog # 516724-500
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :94.11 g/mol
Merck Index:12,07390
Boiling Pt:181.7 °C (1013 hPa)
Melting Pt:40.8 °C
Density:1.0722 g/cm³ (20 °C)
Flash Pt:81 °C
Storage Temperature:Ambient
EINECS:203-632-7
UN:1671
ADR:6.1,II
REACH:01-2119471329-32
 :GHS06
 :GHS05
 :GHS08
CAS Number:108-95-2
Formula:C₆H₅OH
MDL Number:MFCD00002143