

3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) ≥98%, yellow powder cell culture reagent
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:298-93-1
:
Formula:C₁₈H₁₆N₅BrS
MDL Number:MFCD00011964
:414.33 g/mol
Melting Pt:189…199 °C
EINECS:206-069-5
:GHS07
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Identity Test:Passes
- Purity:~98%
- FTIR:Conforms to Standard
- Melting Point:189 - 199°C
- UV/Vis: I max (methanol):376 ± 2 nm; 242 ± 2 nm
Specification Test Results
Frequently Bought Together